Inverted microscope, CKX53
Power supply for mercury vapor lamp 100 W, input voltage 100 - 240 V
Catalog # OLYMN2251000
Supplier: OLYMPUS OPTICAL
Jump to:
Product Details & Documents
Ergonomically designed and enhanced imaging for cell culture, the CKX53 is designed for cell sampling and manipulation, especially for fluorescence observations.
- Observations in lightfield transmission and phase contrast evolutionary to fluorescence
- High contrast thanks to integrated phase system (IPC) (without ring change)
- Three-position fluorescence illuminator: Two sets of filters for excitation (blue and green), empty position, manual shutter and neutral filter slide 6%
Compatible with CKX3-SLP and IX2-SL condenser slides, CKX3-SPLIC, CKX3-SLPAS, IX2-SPLC, IX2-SLPH1, IX2-SLPH2 optical elements, CKX3-MVR object guide, CKX3-HO35DM Petri dish rack, reflective fluorescence illuminator and mercury, metal halide or LED light sources.
: Included CACHN 10× / 0,25 Ph / IPC lens, power cable, platinum insert, Allen key, power supply, two camera cable clips with screws - contrast-inverted 3-D views, optional.
Specifications
- Description:Power supply for mercury vapor lamp 100 W, input voltage 100 - 240 V
Accessories
Product Information
- Catalog #AvailabilityPrice
- ND-25 neutral density filter for transmitted light (43ND-25W45)ND-25 neutral density filter for transmitted light (43ND-25W45)OLYM035137Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- IX2-SLPH1, PH1 Ring For IX2-SL
Product Information
- Catalog #AvailabilityPrice
- Interference filter
Product Information
- Catalog #AvailabilityPrice
- Neutral density filter (50% transmission), 32ND50
Product Information
- Catalog #AvailabilityPrice
- Neutral density filter (12% transmission), 32ND12
Product Information
- Catalog #AvailabilityPrice
- CKX-RCD Relief Contrast Condenser for use with CKX41/. CKX31 microscopesCKX-RCD Relief Contrast Condenser for use with CKX41/. CKX31 microscopesOLYMN1157782Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- CKX3-SLPIC inversion contrast ring slit for CKX53 and IX53CKX3-SLPIC inversion contrast ring slit for CKX53 and IX53OLYMN5228600Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- CKX3-SLPAS aperture stop ring for CKX3-SLP when using PLN2X objectiveCKX3-SLPAS aperture stop ring for CKX3-SLP when using PLN2X objectiveOLYMN5228700Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- Dust Cover for CKX Microscopes
Product Information
- Catalog #AvailabilityPrice
- CK2-SS stage extension plate for CK, CKX and IX microscopesCK2-SS stage extension plate for CK, CKX and IX microscopesOLYM030362Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- Power cable with elbow
Product Information
- Catalog #AvailabilityPrice
- IX2-SL, three position centric phase contrast slider, ring phase stop IX2-PHL and two empty positionsIX2-SL, three position centric phase contrast slider, ring phase stop IX2-PHL and two empty positionsOLYM035793Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- IX2-SLPHC, PHC phase annular stop for center slider position and for CPL10x PH, CPLFL10xPH, LCACH20xPH, for IX2-SL condenser slideIX2-SLPHC, PHC phase annular stop for center slider position and for CPL10x PH, CPLFL10xPH, LCACH20xPH, for IX2-SL condenser slideOLYM035794Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- 45-LBD-IF Daylight Interference Filter
Product Information
- Catalog #AvailabilityPrice
- Neutral density filter (25% transmission), 32ND25
Product Information
- Catalog #AvailabilityPrice
- CKX53SF microscope frame (base body)
Product Information
- Catalog #AvailabilityPrice
- Glass insert for turntables, compatible with 4 × to 20 × objectivesGlass insert for turntables, compatible with 4 × to 20 × objectivesOLYM031540Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- Neutral density filter (6% transmission), 32ND6
Product Information
- Catalog #AvailabilityPrice
- CKX3-HOUN, Universal holder (slides and dishes) for CKX3-MVRCKX3-HOUN, Universal holder (slides and dishes) for CKX3-MVROLYMN5229000Log in for availability1 * 1 Pce - 646,00 €
Product Information
- Catalog #AvailabilityPrice
- IX2-SLPH2, PH2 phase annular stop for right position of IX2-SL condenserIX2-SLPH2, PH2 phase annular stop for right position of IX2-SL condenserOLYM035795Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- CKX3-HO35DM, Insert to hold three 35mm dishes for CKX53 MicroscopeCKX3-HO35DM, Insert to hold three 35mm dishes for CKX53 MicroscopeOLYMN5228900Log in for availability1 * 1 Pce
Product Information
- Catalog #AvailabilityPrice
- CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53OLYMN5228500Log in for availability1 * 1 Pce - 114,00 €



