Order Entry
France
Orders LinkContactUsLinkComponent
Safety Litter Boxes

Safety Litter Boxes

Safety litter box, Stainless steel 18/10, 120 L, 400×300×740 mm
Catalog # BOCH11422
Supplier:   BOCHEM
Safety Litter Boxes
Catalog # BOCH11422
Supplier:  BOCHEM
Safety Litter Boxes
Safety Litter Boxes

Safety litter box, Stainless steel 18/10, 120 L, 400×300×740 mm

Price
737,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents

Made from stainless steel 18/10, electro-polished.

  • With swinging lid
  • With handles left and right
  • Easy to handle and easy to clean
Specifications
  • Capacity:
    120 L
  • Description:
    Safety litter box
  • Material:
    Stainless steel 18/10
  • Seq:
    02
  • W×D×H:
    400×300×740 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.