Order Entry
France
Orders LinkContactUsLinkComponent
Digested lambda DNA
Digested lambda DNA
Lambda DNA digested with Eco RI and Hind III, 20 µG
Catalog # EDVO710
Supplier:   EDVOTEK
Digested lambda DNA
Catalog # EDVO710
Supplier:  EDVOTEK
CAS Number:  
Digested lambda DNA
Lambda DNA digested with Eco RI and Hind III, 20 µG
CAS Number:  
Price
49,50 €
Log in for availability
1 * 20 µG
Product Details & Documents
20 µg for 20 gels.
Specifications
Specifications
  • Pk:
    20 µG
  • Description:
    Lambda DNA digested with Eco RI and Hind III