Wipes for general applications, Tork Premium, colour coded
Blue, 300×380 mm, 8 packs of 40 folded wipes
Catalog # 115-0584
Supplier: Essity
Jump to:
Product Details & Documents
Low Linting, colour coded cloth wipes for cleaning, made from a mixture of viscose and polyester.
- Useful for a colour coded cleaning system
- Tork Premium: Strongest quality for heaviest wiping tasks
Specifications
- Packed:8 packs of 40 folded wipes
- Colour:Blue
- L×W:300×380 mm