Order Entry
France
Orders LinkContactUsLinkComponent
Capillary columns, Chromolith® CapRod®

Capillary columns, Chromolith® CapRod®

Capillary, RP-8e, 0,10 mm, 150 mm
Catalog # 1.50400.0001
Supplier:   Merck
Capillary columns, Chromolith® CapRod®
Catalog # 1.50400.0001
Supplier:  Merck
Capillary columns, Chromolith® CapRod®
Capillary columns, Chromolith® CapRod®

Capillary, RP-8e, 0,10 mm, 150 mm

Price
1 440,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents

Fast nano-LC for proteins and peptides. Chromolith® CapRod® capillaries combine the speed of monolithic silica technology with the sensitivity of nano-LC, thus enabling high speed high sensitivity LC-MS analytics.

  • High flow rates at low pressure
  • Long column lifetime
  • Robust handling
Specifications
  • Column Format:
    Capillary
  • Ø int.:
    0,10 mm
  • Length:
    150 mm
  • Seq:
    02
  • Phase:
    RP-8e
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.