Stoppers, universal caps, FitsAll™
Pale purple, 12 - 16 mm
Catalog # SIMPT403L
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Designed for easy-on, easy-off use. Cap removal is simple and quick. FitsAll™ closure is ideal for recapping most 12 and 16 mm O.D. evacuated glass blood collection tubes. Flexible FitsAll™ closures protect from aerosols of highly infectious microorganisms. They guard samples against cross-contamination and laboratory work areas against infection and spillage. Precision molded from low-density polyethylene, FitsAll™ fits firmly on the tube. Not suitable for autoclaving.
- PE
- Very sturdy under most rigorous procedures, including centrifugation
- Available in 8 colours for easy sample identification
Specifications
- Ø bottom:12 - 16 mm
- Colour:Pale purple