Order Entry
France
Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), VWR®

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), VWR®

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 5 L
Catalog # 87476.360
Supplier:   PROLABO CMP
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), VWR®
Catalog # 87476.360
Supplier:  PROLABO CMP
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), VWR®

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 5 L

CAS Number:  
Price
261,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Specifications

Specifications

  • Pk:
    5 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.