Order Entry
France
Orders LinkContactUsLinkComponent
Safety spectacles, Tracker
Safety spectacles, Tracker
Anti-fog, anti-scratch, Transparent, Clear PC
Catalog # 111-0390
Supplier:   Bollé Safety
Safety spectacles, Tracker
Catalog # 111-0390
Supplier:  Bollé Safety
Safety spectacles, Tracker
Safety spectacles, Tracker
Anti-fog, anti-scratch, Transparent, Clear PC
Price
19,90 €
Log in for availability
1 * 1 Pce
Product Details & Documents
These sporty design safety spectacles feature a durable PC frame, a smoke PC dual lens, straight temples with upper and lower protection and an adjustable headband.
  • Interchangeable foam reinforcement with indirect ventilation
  • Adjustable head strap and rubber arm tips provide a secure fit and greater protection
  • ALS anti-fog/anti-scratch coating on both sides of clear and smoke lenses
  • Rubberised eye seal provides exceptional protection against dust and other particles
Weight: 52 g
EN 166
Certifications: EN 166
Delivery information: Supplied with microfibre pouch.
Specifications
  • Type:
    Anti-fog, anti-scratch
  • Colour:
    Transparent
  • Seq:
    02
  • Lenses:
    Clear PC