Standards for 7100/7500/PT25P + cert 1 set 1 * 1 SET
Catalog # PALIPT804
Supplier: Palintest
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH electrodes, InLab®, Expert Series
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals