Order Entry
France
ContactUsLinkComponent
Ph-combination electrode with sensor recognition, temperature sensor pt 1000, 1 m fixed cable with bnc and banana plug, glass shaft, ceramic diaphragm, electrolyte kcl 3 mol/l, iodine/iodide-reference system, sphere membrane, h-glass, length 170 mm, 1 * 1 items
Catalog # 662-6094
Supplier:  SI Analytics
Ph-combination electrode with sensor recognition, temperature sensor pt 1000, 1 m fixed cable with bnc and banana plug, glass shaft, ceramic diaphragm, electrolyte kcl 3 mol/l, iodine/iodide-reference system, sphere membrane, h-glass, length 170 mm, 1 * 1 items
Catalog # 662-6094
Supplier:  SI Analytics
Frequently Bought Together