Order Entry
France
ContactUsLinkComponent
Vial, capacity 200µl, conical base, material glass, colour clear, diameter 6mm, height 32mm, 8mm crimp top, total volume 250µl, resi dual capacity <2µl, for Perkin Elmer ISS-100, Perkin Elmer ISS-200
Catalog # CRMAC4008-632C
Supplier:  CHROMACOL
Vial, capacity 200µl, conical base, material glass, colour clear, diameter 6mm, height 32mm, 8mm crimp top, total volume 250µl, resi dual capacity <2µl, for Perkin Elmer ISS-100, Perkin Elmer ISS-200
Catalog # CRMAC4008-632C
Supplier:  CHROMACOL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.