Cap, target DP, colour blue, material polypropylene, cap size 9-425, closed top, septum white virgin PTFE 0.01 inches, preferred for longterm sample storage
Catalog # CRMAC4000-99
Supplier: CHROMACOL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.