: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- :123-91-1
- :C₄H₈O₂
- :MFCD00006571
- :88.11 g/mol
- :13,03330
- :101.1 °C (1013 hPa)
- :11.8 °C
- :1.03 g/cm³ (20 °C)
- :12 °C
- :Ambient
- :1165
- :3,II
- :M-Clarity™ quality level = MQ100
- :204-661-8