Order Entry
France
Orders LinkContactUsLinkComponent
Propionic acid, Supelco®

Propionic acid, Supelco®

Glass bottle, 1 mL
Catalog # 94425-1ML-F
Corrosive
Irritant
Supplier:   Merck
Propionic acid, Supelco®
Catalog # 94425-1ML-F
Supplier:  Merck
CAS Number:  79-09-4
Propionic acid, Supelco®
Propionic acid, Supelco®

Glass bottle, 1 mL

CAS Number:  79-09-4
Price
41,60 €
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:0.99 g/cm³ (20 °C)
  • Flash Pt:49 °C
  • UN:3463
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • REACH:01-2119486971-24

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.