Benzophenone
Glass bottle for solids, 5 g
Catalog # XXXX18870
Jump to:
Discontinued
Product Details & Documents
Synonyms: Diphenylmethanone, Diphenyl ketone
- CAS Number:119-61-9
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00003076
- Molecular Weight:182.22 g/mol
- Merck Index:13,01099
- Boiling Pt:305.4 °C (1013 hPa)
- Melting Pt:47…49 °C
- Density:1.085 g/cm³ (20 °C)
- Flash Pt:150 °C
- Storage Temperature:Ambient
- EINECS:204-337-6
- UN:3077
- ADR:9,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



