Cabinet wine gwt4677 storage 462l +5 to +20c 115 bottle capacity 1855mm tall 1 * 1 items
LIEBGWT4677
: Liebherr
Cabinet wine gwt4677 storage 462l +5 to +20c 115 bottle capacity 1855mm tall 1 * 1 items
LIEBGWT4677
: Liebherr
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK