Filters
Filter set HQ SZX2-FGFPHQ, High performance for GFP
630-0958
Filter units, ranging from UV excitation to RFP, respond to applications using various fluorescent dyes and protein.
High quality (HQ) filters have an edge steepness and high transmission that efficiently detect the light of fluorescence to enhance and capture brighter fluorescence images in precise detail.
- For:High performance for GFP
- Description:Filter set HQ SZX2-FGFPHQ
- Seq:05