Filters
Filter set, SZX2-FUV, UV excitation
Catalog # 630-0946
Supplier: OLYMPUS OPTICAL
Jump to:
Product Details & Documents
Filter units, ranging from UV excitation to RFP, respond to applications using various fluorescent dyes and protein.
High quality (HQ) filters have an edge steepness and high transmission that efficiently detect the light of fluorescence to enhance and capture brighter fluorescence images in precise detail.
Specifications
- For:UV excitation
- Description:Filter set, SZX2-FUV
- Seq:09