Order Entry
France
Orders LinkContactUsLinkComponent
Objectives
Objectives
Plan apochromatic objective, 6,2 mm
Catalog # OLYMN1480200
Supplier:   OLYMPUS OPTICAL
Objectives
Catalog # OLYMN1480200
Supplier:  OLYMPUS OPTICAL
Objectives
Objectives
Plan apochromatic objective, 6,2 mm
Price
1 560,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Specifications
  • Magnification:
    2x
  • Description:
    Plan apochromatic objective
  • Seq:
    01
  • Working distance:
    6,2 mm