Order Entry
France
Orders LinkContactUsLinkComponent
Potassium hydroxide ≥85%, pellets

Potassium hydroxide ≥85%, pellets

Glass bottle for solids, 500 g
Catalog # A18854.36
Corrosive
Irritant
Potassium hydroxide ≥85%, pellets
Catalog # A18854.36
CAS Number:  1310-58-3
Potassium hydroxide ≥85%, pellets
Potassium hydroxide ≥85%, pellets

Glass bottle for solids, 500 g

CAS Number:  1310-58-3
Price
38,40 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 85% CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Odour: Odourless pH: 13,5 Melting Point: 360° Boiling Point: 1320-1324° Merck Reference: 14,7640 UN No.: 1813 MDL No.: MFCD00003553

Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Merck Index:14,7640
  • Boiling Pt:1320…1324 °C
  • Melting Pt:360 °C
  • Density:2.044
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1813
  • ADR:8,II
  • REACH:01-2119487136-33
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications