Order Entry
France
Orders LinkContactUsLinkComponent
Adenosine ≥99%

Adenosine ≥99%

Glass bottle for solids, 10 g
Catalog # A10781.09
Adenosine ≥99%
Catalog # A10781.09
CAS Number:  58-61-7
Adenosine ≥99%
Adenosine ≥99%

Glass bottle for solids, 10 g

CAS Number:  58-61-7
Price
47,50 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 99% CAS No.: 58-61-7 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Storage Temperature: 2°C to 8°C Physical Form: Powder Odour: Odourless Melting Point: 234-237° Beilstein Reference: 93029 Merck Reference: 14,153 Optical Rotation: -70° (c=2 in 5% NaOH) MDL No.: MFCD00005752

Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Merck Index:14,153
  • Melting Pt:234…237 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-389-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.