Order Entry
France
Orders LinkContactUsLinkComponent
Silver plating solution ∼2,1 g/l metal content, electroless

Silver plating solution ∼2,1 g/l metal content, electroless

250 mL
Catalog # 44068.AK
Toxic
Silver plating solution ∼2,1 g/l metal content, electroless
Catalog # 44068.AK
CAS Number:  
Silver plating solution ∼2,1 g/l metal content, electroless
Silver plating solution ∼2,1 g/l metal content, electroless

250 mL

CAS Number:  
Price
51,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Physical Form: Liquid Density: 1.04 Appearance: Colourless Melting Point: -5° Boiling Point: 103-105° UN No.: UN3287 MDL No.: MFCD00003397

Applications: For plating on copper, brass or copper alloys

  • MDL Number:MFCD00003397
  • Boiling Pt:103…105 °C
  • Melting Pt:–5 °C
  • Density:1.04
  • Storage Temperature:Ambient
  • UN:3287
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.