Order Entry
France
Orders LinkContactUsLinkComponent
Silver plating solution ∼28,7 g/l (metals basis)

Silver plating solution ∼28,7 g/l (metals basis)

1 L
Catalog # 44067.K2
Toxic
Health
Corrosive
Silver plating solution ∼28,7 g/l (metals basis)
Catalog # 44067.K2
CAS Number:  
Silver plating solution ∼28,7 g/l (metals basis)
Silver plating solution ∼28,7 g/l (metals basis)

1 L

CAS Number:  
Price
182,00 €
Log in for availability
1 * 1 L
Product Details & Documents

UN No.: UN1935 MDL No.: MFCD00003397

  • MDL Number:MFCD00003397
  • Storage Temperature:Ambient
  • UN:1935
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.