Order Entry
France
Orders LinkContactUsLinkComponent
Silver plating solution ∼28,7 g/l (metals basis)

Silver plating solution ∼28,7 g/l (metals basis)

250 mL
Catalog # 44067.AK
Toxic
Health
Corrosive
Silver plating solution ∼28,7 g/l (metals basis)
Catalog # 44067.AK
CAS Number:  
Silver plating solution ∼28,7 g/l (metals basis)
Silver plating solution ∼28,7 g/l (metals basis)

250 mL

CAS Number:  
Price
62,90 €
Log in for availability
1 * 250 mL
Product Details & Documents

UN No.: UN1935 MDL No.: MFCD00003397

  • MDL Number:MFCD00003397
  • Storage Temperature:Ambient
  • UN:1935
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.