Aluminium oxide, regular drying agent
Glass bottle for solids, 2 kg
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 3.2mm Spheres Odour: Odourless MDL No.: MFCD00003424
Dry gases and liquids. Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2. Liquids include: benzene, hexane, oil, pentane.
: Aluminium(III) oxide, Alumina C, Aluminum oxide
- :1344-28-1
- :Al₂O₃
- :MFCD00003424
- :101.96 g/mol
- :13,00355
- :2980 °C (1013 hPa)
- :2045 °C
- :3.94 g/cm³ (20 °C)
- :Ambient
- :215-691-6
- :01-2119529248-35
- Pack type:Glass bottle for solids
- Pk:2 kg



