Aluminium oxide, regular drying agent
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Aluminium oxide, regular drying agent
Catalog # 44018.22
Supplier: Thermo Fisher Scientific
CAS Number: 1344-28-1
Jump to:
Product Details & Documents
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 3.2mm Spheres Odour: Odourless MDL No.: MFCD00003424
Dry gases and liquids. Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2. Liquids include: benzene, hexane, oil, pentane.
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
- CAS Number:1344-28-1
- Formula:Al₂O₃
- MDL Number:MFCD00003424
- Molecular Weight:101.96 g/mol
- Merck Index:13,00355
- Boiling Pt:2980 °C (1013 hPa)
- Melting Pt:2045 °C
- Density:3.94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-691-6
- REACH:01-2119529248-35
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



