Dynamometers
LG-50KG, 0 - 50 kgf, 0,25 kgf
Catalog # LLOYLG50KG
Supplier: LLOYD INSTRUMENTS
Jump to:
Product Details & Documents
The Chatillon® LG series mechanical force gauge is ideal for a wide range of force testing applications. Features a larger dial for improved resolution and readability, peak hold button, ergonomic shape and improved accuracy of +0,5% of full scale.
- Graduation styles : KGF and Newtons
- Excellent, economical choice for tensile or compression testing
- NIST traceable calibration certificate
Specifications
- Range:0 - 50 kgf
- Description:LG-50KG
- Seq:06
- Readability:0,25 kgf