Order Entry
France
ContactUsLinkComponent
Urine cup 100ml with paper cover pp,urine cup - polypropylene (pp) 100 ml autoclaved up to +121c self-adhesive, tight-fitting lid a nd paper inscribable. 1 * 1.000 items
  SEMA2038
 :  SEMADENI
Urine cup 100ml with paper cover pp,urine cup - polypropylene (pp) 100 ml autoclaved up to +121c self-adhesive, tight-fitting lid a nd paper inscribable. 1 * 1.000 items
  SEMA2038
 :  SEMADENI
Frequently Bought Together