Order Entry
France
Orders LinkContactUsLinkComponent
 

Ethanol 90% (v/v), TechniSolv®, pure

Plastic container, 5 L
Catalog # 83811.360
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Ethanol 90% (v/v), TechniSolv®, pure
Catalog # 83811.360
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  64-17-5
Ethanol 90% (v/v), TechniSolv®, pure
Ethanol 90% (v/v), TechniSolv®, pure

Plastic container, 5 L

CAS Number:  64-17-5
Price
71,10 €
Log in for availability
1 * 5 L
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

 : WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
Specifications

Specification Test Results

  • Assay:
    Min. 89.5 %
  • Density (20/20):
    0.827 - 0.833 kg/l

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Cat. No.:
    83811.360
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.