Order Entry
France
Orders LinkContactUsLinkComponent
Ammonia solution 25% 100 ml/l solution

Ammonia solution 25% 100 ml/l solution

Plastic bottle, 5 L
Catalog # PRLS85163.360
Irritant
Environment
Supplier:   PROLABO CMP
Ammonia solution 25% 100 ml/l solution
Catalog # PRLS85163.360
Supplier:  PROLABO CMP
CAS Number:  1336-21-6
Ammonia solution 25% 100 ml/l solution

Plastic bottle, 5 L

CAS Number:  1336-21-6
Price
112,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Concentrated solution to dilute to 100 mL/L.

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications