Order Entry
France
ContactUsLinkComponent
Unique two-yarn system for fabric construction incorporates a microdenier yarn and a regular polyester yarn to create a wiper with s uperior sorption and wiping characteristics. Both yarns are 100% continuous filament polyester engineered with Milli
Catalog # 115-0441
Supplier:  CONTEC
Unique two-yarn system for fabric construction incorporates a microdenier yarn and a regular polyester yarn to create a wiper with s uperior sorption and wiping characteristics. Both yarns are 100% continuous filament polyester engineered with Milli
Catalog # 115-0441
Supplier:  CONTEC