Order Entry
France
ContactUsLinkComponent
Laboratory pH meter Lab 850 Set. Measuring parameters pH, mV, temp., microprocessor, DIN 19262 connection. Including cover Z 880, st and S4D Z 865, power supply Z 850, pH-temp. combination electrode BlueLine 14 pH, calibration solutions (DIN). 1 * 1 items
Catalog # 663-0086
Supplier:  SI Analytics
undefined
Laboratory pH meter Lab 850 Set. Measuring parameters pH, mV, temp., microprocessor, DIN 19262 connection. Including cover Z 880, st and S4D Z 865, power supply Z 850, pH-temp. combination electrode BlueLine 14 pH, calibration solutions (DIN). 1 * 1 items
Catalog # 663-0086
Supplier:  SI Analytics
Frequently Bought Together