Order Entry
France
Orders LinkContactUsLinkComponent
Nitromethane ≥98%

Nitromethane ≥98%

Glass bottle for solids, 500 g
Catalog # A11806.36
Flammable
Irritant
Nitromethane ≥98%
Catalog # A11806.36
CAS Number:  75-52-5
Nitromethane ≥98%
Nitromethane ≥98%

Glass bottle for solids, 500 g

CAS Number:  75-52-5
Price
79,00 €
Log in for availability
1 * 500 g
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98+% CAS No.: 75-52-5 Molecular Formula: CH3NO2 Formula Weight: 61.04 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.130 Refractive Index: 1.3820 Appearance: Colourless Odour: Characteristic Melting Point: -29° Boiling Point: 101° Beilstein Reference: 1698205 Merck Reference: 14,6611 UN No.: 1261 MDL No.: MFCD00007400

  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:14,6611
  • Boiling Pt:101 °C
  • Melting Pt:–29 °C
  • Density:1.130
  • Flash Pt:35 °C (95 °F)
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.