Order Entry
France
Orders LinkContactUsLinkComponent
 

Amiloride, Reference Standards

Amiloride Hydrochloride, USP reference standard, Vial, 500 mg
  1019701.
 :   USP
Amiloride, Reference Standards
  1019701.
 :  USP
 :  
Amiloride, Reference Standards

Amiloride Hydrochloride, USP reference standard, Vial, 500 mg

 :  
 
397,00 €
   
1 * 500 mg
 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Pack type:
    Vial
  • Pk:
    500 mg
  • Standards:
    USP reference standard
  • Description:
    Amiloride Hydrochloride
  • UN No.:
    2811
  • Cat. No.:
    1019701.
  • Formula:
    C₆H₈ClN₇O·HCl·₂H₂O
  • Description:
    Amiloride Hydrochloride
  • CAS No.:
    17440-83-4