Order Entry
France
Orders LinkContactUsLinkComponent
Sumatriptan Related Compounds, Reference Standard

Sumatriptan Related Compounds, Reference Standard

Sumatriptan Succinate Related Compound C, USP reference standard, Vial, 50 mg
Catalog # 1642223.
Supplier:   USP
Sumatriptan Related Compounds, Reference Standard
Catalog # 1642223.
Supplier:  USP
CAS Number:  
Sumatriptan Related Compounds, Reference Standard

Sumatriptan Succinate Related Compound C, USP reference standard, Vial, 50 mg

CAS Number:  
Price
1 620,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    USP reference standard
  • Description:
    Sumatriptan Succinate Related Compound C
  • Cat. No.:
    1642223.
  • Description:
    Sumatriptan Succinate Related Compound C
  • Synonym(s):
    [3-[2-(dimethylamino)ethyl]-1-(hydroxymethyl)-1H-indol-5-yl]-N-methylmethanesulfonamide succinate salt

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.