Order Entry
France
Orders LinkContactUsLinkComponent
Dexamethasone, Reference Standards
Dexamethasone Acetate, USP reference standard, Vial, 200 mg
  1176506.
 :   USP
Dexamethasone, Reference Standards
  1176506.
 :  USP
 :  
Dexamethasone, Reference Standards
Dexamethasone Acetate, USP reference standard, Vial, 200 mg
 :  
405,00 €
1 * 200 mg
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard
  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Dexamethasone Acetate
  • Cat. No.:
    1176506.
  • Description:
    Dexamethasone Acetate
  • CAS No.:
    1177-87-3