Order Entry
France
Orders LinkContactUsLinkComponent
Flash point tester reference materials

Flash point tester reference materials

Flash Check, secondary verification material, 3
Catalog # 751-0029
Supplier:   STANHOPE SETA
Flash point tester reference materials
Catalog # 751-0029
Supplier:  STANHOPE SETA
CAS Number:  
Flash point tester reference materials
Flash point tester reference materials

Flash Check, secondary verification material, 3

CAS Number:  
Price
660,00 €
Log in for availability
1 * 3 Pce
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifications

Specifications

  • Pk:
    3
  • Description:
    Flash Check, secondary verification material

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.