Order Entry
France
Orders LinkContactUsLinkComponent
Push-in stoppers for tubes

Push-in stoppers for tubes

Green, 17 mm
Catalog # SIMPT401-10G
Supplier:   Simport Scientific
Push-in stoppers for tubes
Catalog # SIMPT401-10G
Supplier:  Simport Scientific
Push-in stoppers for tubes
Push-in stoppers for tubes

Green, 17 mm

Price
80,60 €
Log in for availability
1 * 1.000 Pce
Product Details & Documents

HDPE. Fit tubes of 15 ml capacity and 17 mm Ø.

  • Impermeable to liquids
  • Can be inserted in tubes and removed again easily with one hand
  • Economical, can also be used as disposable stoppers
Specifications
  • Ø bottom:
    17 mm
  • Colour:
    Green
  • Seq:
    12
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.