Sealing Rings 1 * 1 items
Catalog # METR6.1244.040
Supplier: METROHM
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
uvex pheos Safety Spectacles
Supplier: UVEX
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Rotary evaporating flasks
Supplier: Lenz Laborglas GmbH & CO.KG
Phenolphthalein 98.0-101.0% Ph. Eur.
Supplier: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Glass microfibre filters without binder, grade GF/C, Whatman™
Supplier: Whatman products (Cytiva)
Bag closures, WeLoc®
Supplier: WELOC
Nitric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Accessories for PURELAB Flex Systems
Supplier: ELGA LabWater