Aluminium oxide, intermediate surface area catalyst support
Glass bottle for solids, 5 kg
Supplier: Thermo Fisher Scientific
Aluminium oxide, intermediate surface area catalyst support
Catalog # 43856.A7
Supplier: Thermo Fisher Scientific
CAS Number: 1344-28-1
Jump to:
Product Details & Documents
Physical Form: 1/8" pellets MDL No.: MFCD00003424
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
- CAS Number:1344-28-1
- Formula:Al₂O₃
- MDL Number:MFCD00003424
- Molecular Weight:101.96 g/mol
- Merck Index:13,00355
- Boiling Pt:2980 °C (1013 hPa)
- Melting Pt:2045 °C
- Density:3.94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-691-6
- REACH:01-2119529248-35
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



