Aluminium oxide, intermediate surface area catalyst support
Glass bottle for solids, 1 kg
Aluminium oxide, intermediate surface area catalyst support
43856.A1
: 1344-28-1
Physical Form: 1/8" pellets MDL No.: MFCD00003424
: Aluminium(III) oxide, Alumina C, Aluminum oxide
- :1344-28-1
- :Al₂O₃
- :MFCD00003424
- :101.96 g/mol
- :13,00355
- :2980 °C (1013 hPa)
- :2045 °C
- :3.94 g/cm³ (20 °C)
- :Ambient
- :215-691-6
- :01-2119529248-35
- Pack type:Glass bottle for solids
- Pk:1 kg