Agarose
Glass bottle for solids, 1 kg
Catalog # TREV9700-01K-P
Marketsource
Supplier: TREVIGEN
Aller à :
Product Details & Documents
- Numéro CAS:9012-36-6
- Numéro fabricant:MFCD00081294
- Point de fusion:60…90 °C
- Température de stockage:Ambient
- EINECS:232-731-8
Spécifications
Spécifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



