Order Entry
France
Orders LinkContactUsLinkComponent
Flash point tester reference materials
Small scale certified Flash point material ( closed cups), nominal flash point 74 °C, 50 mL
  STAN99878-3
Flash point tester reference materials
  STAN99878-3
 :  
Flash point tester reference materials
Flash point tester reference materials
Small scale certified Flash point material ( closed cups), nominal flash point 74 °C, 50 mL
 :  
601,00 €
1 * 50 mL
  • Pk:
    50 mL
  • Description:
    Small scale certified Flash point material ( closed cups), nominal flash point 74 °C