Order Entry
France
ContactUsLinkComponent
Conductivity cell LF 5100+ with SMEK plug head. 2 pole cell, plastic shaft, sensor 2 platinum rings, cell constant 1.0 cm-1, length 120 mm, 12 mm , -5...+80 C 1 * 1 items
Catalog # 663-0276
Supplier:  SI Analytics
Conductivity cell LF 5100+ with SMEK plug head. 2 pole cell, plastic shaft, sensor 2 platinum rings, cell constant 1.0 cm-1, length 120 mm, 12 mm , -5...+80 C 1 * 1 items
Catalog # 663-0276
Supplier:  SI Analytics
Frequently Bought Together