ASTM colour 5 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0043
Supplier: Lovibond Tintometer
ASTM colour 5 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0043
Supplier: Lovibond Tintometer
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Certified colour reference standards
Supplier: Lovibond Tintometer