Order Entry
France
Orders LinkContactUsLinkComponent
Sample tubes

Sample tubes

2,0 ml, Free standing, Clear, with side label, graduated in 0,25 ml divisions, PP, screw cap with attachment loop, Sterile
Catalog # BZYM710067
Marketsource
Supplier:   BIOZYM
Sample tubes
Catalog # BZYM710067
Supplier:  BIOZYM
Marketsource
Sample tubes

2,0 ml, Free standing, Clear, with side label, graduated in 0,25 ml divisions, PP, screw cap with attachment loop, Sterile

Price
Inquire For Price
Log in for availability
1 * 500 Pce
Product Details & Documents

PP, translucent.

  • Excellent chemical resistance
  • Excellent temperature resistance

With or without screw caps.

Specifications
  • Sterility:
    Sterile
  • Type:
    screw cap with attachment loop
  • Capacity:
    2,0 ml
  • Colour:
    Clear
  • Description:
    with side label, graduated in 0,25 ml divisions
  • Material:
    PP
  • Seq:
    24
  • Base:
    Free standing
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.