Order Entry
France
Orders LinkContactUsLinkComponent
Sample tubes
Sample tubes
1,5 ml, Free standing, Clear, with side label, PP, screw cap with attachment loop, Sterile
Catalog # BZYM710066
Marketsource
Supplier:   BIOZYM
Sample tubes
Catalog # BZYM710066
Supplier:  BIOZYM
Marketsource
Sample tubes
1,5 ml, Free standing, Clear, with side label, PP, screw cap with attachment loop, Sterile
Price
Inquire For Price
Log in for availability
1 * 500 Pce
Product Details & Documents
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
With or without screw caps.
Specifications
  • Sterility:
    Sterile
  • Type:
    screw cap with attachment loop
  • Capacity:
    1,5 ml
  • Colour:
    Clear
  • Description:
    with side label
  • Material:
    PP
  • Seq:
    20
  • Base:
    Free standing