Sample tubes
1,5 ml, Conical bottom, Clear, with side label, PP, screw cap with attachment loop, Sterile
Catalog # BZYM710062
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifications
- Sterility:Sterile
- Type:screw cap with attachment loop
- Capacity:1,5 ml
- Colour:Clear
- Description:with side label
- Material:PP
- Seq:19
- Base:Conical bottom