Order Entry
France
Orders LinkContactUsLinkComponent
Sample tubes
Sample tubes
0,5 ml, Conical bottom, Clear, without side label, PP, screw cap with attachment loop, Sterile
  BZYM710061
Marketsource
 :   BIOZYM
Sample tubes
  BZYM710061
 :  BIOZYM
Marketsource
Sample tubes
0,5 ml, Conical bottom, Clear, without side label, PP, screw cap with attachment loop, Sterile
1 * 500 Pce
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
With or without screw caps.
  • Sterility:
    Sterile
  • Type:
    screw cap with attachment loop
  • Capacity:
    0,5 ml
  • Colour:
    Clear
  • Description:
    without side label
  • Material:
    PP
  • Seq:
    15
  • Base:
    Conical bottom