Sample tubes
0,5 ml, Conical bottom, Clear, without side label, PP, screw cap without attachment loop, Sterile
Catalog # BZYM710051
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifications
- Sterility:Sterile
- Type:screw cap without attachment loop
- Capacity:0,5 ml
- Colour:Clear
- Description:without side label
- Material:PP
- Seq:13
- Base:Conical bottom